missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenylsulfonyl Chloride, TRC

Product Code. 30371906
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30371906 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371906 Supplier Toronto Research Chemicals Supplier No. TRCD4268201G
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1656-44-6
Molecular Formula C6H3ClN2O6S
Quantity 1 g
Synonym 2,4-Dinitrobenzene-1-sulfonyl Chloride, 2,4-Dinitrobenzenesulfonyl Chloride
InChI Formula InChI=1S/C6H3ClN2O6S/c7-16(14,15)6-2-1-4(8(10)11)3-5(6)9(12)13/h1-3H
SMILES [O-][N+](=O)c1ccc(c(c1)[N+](=O)[O-])S(=O)(=O)Cl
IUPAC Name 2,4-dinitrobenzenesulfonyl chloride
Molecular Weight (g/mol) 266.62
Formula Weight 265.94
Chemical Name or Material 2,4-Dinitrophenylsulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.