missing translation for 'onlineSavingsMsg'
Learn More

2,4-Disulfamyl-5-trifluoromethylaniline, TRC

Product Code. 30135441
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135441 10 g 10g
30134616 5 g 5g
30008334 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135441 Supplier Toronto Research Chemicals Supplier No. TRCD49358010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654-62-6
Molecular Formula C7 H8 F3 N3 O4 S2
Molecular Weight (g/mol) 319.28
Synonym 4-Amino-6-(trifluoromethyl)benzene-1,3-disulphonamide, 1,3-Benzenedisulfonamide, 4-amino-6-(trifluoromethyl)-, Toluene-2,4-disulfonamide, 5-amino-α,α,α-trifluoro- (6CI,7CI,8CI), 2,4-Disulfamoyl-5-trifluoromethylaniline, 4-Amino-6-(trifluoromethyl)-1,3-benzenedisulfonamide, 5-Trifluoromethyl-2,4-disulfamoylaniline, NSC 44625
IUPAC Name 4-amino-6-(trifluoromethyl)benzene-1,3-disulfonamide
SMILES Nc1cc(c(cc1S(=O)(=O)N)S(=O)(=O)N)C(F)(F)F

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H8F3N3O4S2/c8-7(9,10)3-1-4(11)6(19(13,16)17)2-5(3)18(12,14)15/h1-2H,11H2,(H2,12,14,15)(H2,13,16,17)
Formula Weight 318.9908
Chemical Name or Material 4-amino-6-(trifluoromethyl)benzene-1,3-disulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.