missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)ethanol, 97%

Product Code. 11357515
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357515 5 g 5g
11367515 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357515 Supplier Thermo Scientific Alfa Aesar Supplier No. L05618.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7589-27-7 | C8H9FO | 140.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7589-27-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00002898
InChI Key MWUVGXCUHWKQJE-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl ethanol, 4-fluorophenethyl alcohol, 4-fluorophenylethanol, benzeneethanol, 4-fluoro, 2-4-fluorophenyl ethan-1-ol, 4-fluorophenethylic alcohol, phenethyl alcohol, p-fluoro, 4-fluorobenzeneethanol, 4fa
PubChem CID 82068
IUPAC Name 2-(4-fluorophenyl)ethanol
SMILES OCCC1=CC=C(F)C=C1

Specifications

Density 1.121
Boiling Point 118°C (20 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.5065
Quantity 5 g
Beilstein 2354310
Formula Weight 140.16
Percent Purity 97%
Chemical Name or Material 2-(4-Fluorophenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.