missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Hydroxyphenyl)ethanol, 98%

Product Code. 11482687
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482687 5 g 5g
11472687 1 g 1g
11492687 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482687 Supplier Thermo Scientific Alfa Aesar Supplier No. A14891.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 501-94-0 | C8H10O2 | 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-94-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002902
InChI Key YCCILVSKPBXVIP-UHFFFAOYSA-N
Synonym tyrosol, 2-4-hydroxyphenyl ethanol, 4-hydroxyphenethyl alcohol, 4-2-hydroxyethyl phenol, 4-hydroxyphenylethanol, 4-hydroxybenzeneethanol, p-hydroxyphenethyl alcohol, benzeneethanol, 4-hydroxy, p-tyrosol, p-hydroxyphenylethanol
PubChem CID 10393
ChEBI CHEBI:1879
IUPAC Name 4-(2-hydroxyethyl)phenol
SMILES C1=CC(=CC=C1CCO)O

Specifications

Melting Point 89°C to 92°C
Boiling Point 139°C (1 mmHg)
Quantity 5 g
Beilstein 1859884
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 2-(4-Hydroxyphenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.