missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Isobutylphenyl)propanenitrile, 97%, Thermo Scientific™

Product Code. 10620933
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10620933 1 g 1g
10526435 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10620933 Supplier Thermo Scientific Maybridge Supplier No. SEW06499DA
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58609-73-7
Molecular Formula C13H17N
Molecular Weight (g/mol) 187.286
MDL Number MFCD02180706
InChI Key PKQKHWNHCKNYSW-UHFFFAOYSA-N
Synonym 2-4-isobutylphenyl propanenitrile, 2-4-isobutylphenyl propionitrile, 2-4-2-methylpropyl phenyl propanenitrile, 2-4-isobutylphenyl propiononitrile, 2-p-isobutylphenyl propiononitrile, +--2-4'-isobutylphenyl propionitrile, +/--2-4'-isobutylphenyl propionitrile, ibuprofen nitrile, aronis24266, 2-4-isobutylphenyl-propionitrile
PubChem CID 108374
ChEBI CHEBI:27415
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanenitrile
SMILES CC(C)CC1=CC=C(C=C1)C(C)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)2CHCH2C6H4CH(CN)CH3
Quantity 1 g
Formula Weight 187.28
Percent Purity 97%
Chemical Name or Material 2-(4-Isobutylphenyl)propanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.