accessibility menu, dialog, popup

UserName

CH3H3CH3CN

CAS RN 58609-73-7

IUPAC Name: 2-[4-(2-methylpropyl)phenyl]propanenitrile
Synonyms: 2-[4-(2-methylpropyl)phenyl]propanenitrile
Molecular Weight (g/mol): 187.29
Molecular Formula: C13H17N
InChi Key: PKQKHWNHCKNYSW-UHFFFAOYNA-N
SMILES: CC(C)CC1=CC=C(C=C1)C(C)C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results