missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenoxy)ethylamine, Thermo Scientific™

Product Code. 10283182
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10283182 250 mg Amber Glass Bottle 250mg
10495902 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10283182

Brand: Thermo Scientific Maybridge RF03346CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50800-92-5
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052978
InChI Key YXRCDWIZAGWUBL-UHFFFAOYSA-N
Synonym 2-4-methoxyphenoxy ethylamine, 2-4-methoxyphenoxy ethanamine, 2-4-methoxyphenoxy-1-ethanamine, 1-2-aminoethoxy-4-methoxybenzene, 2-4-methoxy-phenoxy-ethylamine, 2-4-methoxyphenoxy ethan-1-amine, ethanamine, 2-4-methoxyphenoxy, maybridge3_000601, 2-4-methoxy-phenoxy ethylamine
PubChem CID 1797793
IUPAC Name 2-(4-methoxyphenoxy)ethanamine
SMILES COC1=CC=C(C=C1)OCCN

Specifications

Packaging Amber Glass Bottle
Linear Formula NH2(CH2)2OC6H4OCH3
Quantity 250 mg
Formula Weight 167.21
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-(4-Methoxyphenoxy)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.