missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)acetaldehyde, TRC

Product Code. 30474806
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474806 50 mg 50mg
30480276 100 mg 100mg
30481926 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474806 Supplier Toronto Research Chemicals Supplier No. TRCM22298350MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5703-26-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.17
IUPAC Name 2-(4-methoxyphenyl)acetaldehyde
SMILES COc1ccc(CC=O)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H10O2/c1-11-9-4-2-8(3-5-9)6-7-10/h2-5,7H,6H2,1H3
Formula Weight 150.0681
Chemical Name or Material 2-(4-Methoxyphenyl)acetaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.