missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)ethanol, 98%

Product Code. 11392396
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392396 50 g 50g
11382396 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392396 Supplier Thermo Scientific Alfa Aesar Supplier No. L06158.18
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 702-23-8 | C9H12O2 | 152.193 g/mol

2-(4-Methoxyphenyl)ethanol is used as an internal standard in the fluorous biphasic catalysis reaction. It is used in the preparation of 4-(2-iodoethyl)phenol, by refluxing it with 47% hydriodic acid. It may be used in the preparation of (2R*,4R*)-1-n-butyl-2-methyl-4-(2-oxopyrrolidin-1-yl)-6-methoxy-1,2,3,4-tetrahydroquinoline and (2R*,4S*)-1-n-butyl-2-methyl-4-(2-oxopyrrolidin-1-yl)-6-methoxy-1,2,3,4- tetrahydroquinoline. It is used for foods, perfumes, and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Methoxyphenyl)ethanol is used as an internal standard in the fluorous biphasic catalysis reaction. It is used in the preparation of 4-(2-iodoethyl)phenol, by refluxing it with 47% hydriodic acid. It may be used in the preparation of (2R*,4R*)-1-n-butyl-2-methyl-4-(2-oxopyrrolidin-1-yl)-6-methoxy-1,2,3,4-tetrahydroquinoline and (2R*,4S*)-1-n-butyl-2-methyl-4-(2-oxopyrrolidin-1-yl)-6-methoxy-1,2,3,4- tetrahydroquinoline. It is used for foods, perfumes, and cosmetics.

Solubility
Soluble in DMSO, Methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 702-23-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002900
InChI Key AUWDOZOUJWEPBA-UHFFFAOYSA-N
Synonym 2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol
PubChem CID 69705
IUPAC Name 2-(4-methoxyphenyl)ethanol
SMILES COC1=CC=C(C=C1)CCO

Specifications

Melting Point 26°C to 30°C
Boiling Point 334°C to 336°C
Flash Point >110°C (230°F)
Refractive Index 1.537
Quantity 50 g
Beilstein 2043563
Solubility Information Soluble in DMSO,Methanol.
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-(4-Methoxyphenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.