missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methyl-5-thiazolyl)ethyl acetate, 98+%

Product Code. 11444927
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444927 5 g 5g
11454927 25 g 25g
11464927 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444927 Supplier Thermo Scientific Alfa Aesar Supplier No. B21867.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 656-53-1 | C8H11NO2S | 185.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 656-53-1
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.24
MDL Number MFCD00005338
InChI Key CRTCWNPLKVVXIX-UHFFFAOYSA-N
Synonym 2-4-methylthiazol-5-yl ethyl acetate, 4-methyl-5-thiazolylethyl acetate, 2-4-methyl-5-thiazolyl ethyl acetate, sulfurol acetate, 4-methyl-5-2-acetoxyethyl thiazole, 4-methyl-5-thiazoleethanol acetate, 5-2-acetoxyethyl-4-methylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethyl acetate, 5-thiazoleethanol, 4-methyl-, acetate ester, 4-methyl-5-beta-acetoxyethyl thiazole
PubChem CID 61192
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethyl acetate
SMILES CC(=O)OCCC1=C(C)N=CS1

Specifications

Density 1.147
Boiling Point 116°C to 118°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.51
Quantity 5 g
Formula Weight 185.24
Percent Purity ≥98%
Chemical Name or Material 2-(4-Methyl-5-thiazolyl)ethyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.