missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylpiperazin-1-yl)benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10229432
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10229432 250 mg Amber Glass Bottle 250mg
10749583 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10229432

Brand: Thermo Scientific Maybridge CC36001CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 159589-70-5
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD01788118
InChI Key WKGFDTBUUBBWJZ-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-yl benzoic acid, 2-4-methyl-piperazin-1-yl-benzoic acid, 2-4-methyl-1-piperazinyl benzoic acid, 1-2-carboxyphenyl-4-methylpiperazine, 2-4-methylpiperazinyl benzoic acid, 4-methyl-piperazin-1-yl-benzoic acid, 2-4-methylpiperazin-1-yl benzoicacid, benzoic acid,2-4-methyl-1-piperazinyl, benzoic acid, 2-4-methyl-1-piperazinyl
PubChem CID 1120459
IUPAC Name 2-(4-methylpiperazin-1-yl)benzoic acid
SMILES CN1CCN(CC1)C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 2-(4-Methylpiperazin-1-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.