missing translation for 'onlineSavingsMsg'
Learn More

{2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine, 97%, Thermo Scientific™

Product Code. 10431704
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10431704 250 mg Amber Glass Bottle 250mg
10042303 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431704 Supplier Thermo Scientific Maybridge Supplier No. CC47413CB
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-50-1
Molecular Formula C13H21N3
Molecular Weight (g/mol) 219.332
MDL Number MFCD06797802
InChI Key OKGWICVOLZFMPE-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-ylmethyl benzylamine, 1-2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazin-1-yl methyl phenyl methanamine, 2-4-methylpiperazinyl methyl phenyl methylamine, 2-4-methyl-1-piperazinyl methyl-benzenemethanamine, 2-4-methylpiperazin-1-yl methyl phenyl methylamine, benzenemethanamine,2-4-methyl-1-piperazinyl methyl
PubChem CID 16228064
IUPAC Name [2-[(4-methylpiperazin-1-yl)methyl]phenyl]methanamine
SMILES CN1CCN(CC1)CC2=CC=CC=C2CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.33
Percent Purity 97%
Chemical Name or Material {2-[(4-Methylpiperazin-1-yl)methyl]phenyl}methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.