missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Pentanediol, (+/-) + meso, 99%
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.
Solubility
Fully miscible.
Notes
Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.
2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.
Solubility
Fully miscible.
Notes
Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Density | 0.955 |
| Boiling Point | 200°C to 201°C |
| Flash Point | 101°C (213°F) |
| Odor | Odorless |
| Refractive Index | 1.435 |
| Quantity | 10 g |
| Beilstein | 969186 |
| Sensitivity | Hygroscopic |
| Solubility Information | Fully miscible. |
| Formula Weight | 104.15 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction