missing translation for 'onlineSavingsMsg'
Learn More

2-(4-tert-Butylphenyl)ethanol, 96%

Product Code. 11323177
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323177 5 g 5g
11333177 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323177 Supplier Thermo Scientific Alfa Aesar Supplier No. L13611.06
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5406-86-0 | C12H18O | 178.275 g/mol

2-(4-tert-Butylphenyl)ethanol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-tert-Butylphenyl)ethanol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5406-86-0
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00236023
InChI Key NZGMMENPUKHODD-UHFFFAOYSA-N
Synonym 2-4-tert-butylphenyl ethanol, 2-4-tert-butyl phenyl ethanol, p-tert-butylphenethyl alcohol, p-tert-butyl phenethyl alcohol, phenethyl alcohol, p-tert-butyl, 4-tert-butylphenethyl alcohol, unii-aes74i7nru, aes74i7nru, benzeneethanol, 4-1,1-dimethylethyl
PubChem CID 79410
IUPAC Name 2-(4-tert-butylphenyl)ethanol
SMILES CC(C)(C)C1=CC=C(C=C1)CCO

Specifications

Melting Point 31°C to 35°C
Density 0.975
Boiling Point 141°C to 143°C (15 mmHg)
Refractive Index 1.5209
Quantity 5 g
UN Number UN3077
Beilstein 2246595
Solubility Information Insoluble in water.
Formula Weight 178.28
Percent Purity 96%
Chemical Name or Material 2-(4-tert-Butylphenyl)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.