missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Trifluoromethyl)phenyl]ethylamine, 98%

Product Code. 15421619
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421619 5 g 5g
15431619 250 mg 250mg
15411619 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421619 Supplier Thermo Scientific Alfa Aesar Supplier No. H64415.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 775-00-8 | C9H10F3N | 189.181 g/mol

It is applied as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 775-00-8
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD01529875
InChI Key HMFOBPNVAAAACP-UHFFFAOYSA-N
Synonym 4-trifluoromethylphenethylamine, 2-4-trifluoromethyl-phenyl-ethylamine, 2-4-trifluoromethyl phenyl ethanamine, 4-trifluoromethyl phenethylamine, 2-4-trifluoromethyl-phenyl ethylamine, 2-4-trifluoromethylphenyl ethylamine, 2-4-trifluoromethyl phenyl ethan-1-amine, 2-4-trifluoromethyl phenyl ethylamine, 2-4-trifluoromethyl phenyl ethyl amine
PubChem CID 1988106
IUPAC Name 2-[4-(trifluoromethyl)phenyl]ethanamine
SMILES C1=CC(=CC=C1CCN)C(F)(F)F

Spezifikation

Quantity 5 g
UN Number UN2735
Beilstein 2719818
Solubility Information Slightly soluble in water.
Formula Weight 189.18
Percent Purity 98%
Chemical Name or Material 2-[4-(Trifluoromethyl)phenyl]ethylamine

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.