missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(trifluoromethyl)aniline, 99%

Product Code. 11363646
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363646 1 g 1g
11373646 5 g 5g
11383646 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363646 Supplier Thermo Scientific Alfa Aesar Supplier No. L14706.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 328-93-8 | C8H5F6N | 229.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-93-8
Molecular Formula C8H5F6N
Molecular Weight (g/mol) 229.13
MDL Number MFCD00074940
InChI Key XWMVIJUAZAEWIE-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl aniline, 2,5-di trifluoromethyl aniline, 2,5-bis-trifluoromethyl aniline, 2,5-bis-trifluoromethylaniline, benzenamine, 2,5-bis trifluoromethyl, 2,5-bis trifluoromethyl-aniline, 2,5-bis trifluoromethyl benzenamine, 1-amino-2,5-bis trifluoromethyl benzene, 2,5-bis trifluoromethyl phenylamine, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-2,5-xylidine
PubChem CID 719818
IUPAC Name 2,5-bis(trifluoromethyl)aniline
SMILES NC1=CC(=CC=C1C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 70°C to 71°C (15 mmHg)
Flash Point 71°C (159°F)
Odor Recognizable
Quantity 1 g
UN Number UN2810
Beilstein 2653046
Formula Weight 229.13
Percent Purity 99%
Chemical Name or Material 2,5-Bis(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.