missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothiophene, 95%

Product Code. 10502931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10502931 25 g Glass bottle 25g
10041580 100 g Glass bottle 100g
10666584 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10502931 Supplier Thermo Scientific Acros Supplier No. 148680250
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3141-27-3 | C4H2Br2S | 241.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3141-27-3
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.93
MDL Number MFCD00005420
InChI Key KBVDUUXRXJTAJC-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
PubChem CID 18453
IUPAC Name 2,5-dibromothiophene
SMILES BrC1=CC=C(Br)S1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -6°C
Color Yellow to Brown
Density 2.1400g/mL
Boiling Point 211°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.6270 to 1.63
Quantity 25 g
Beilstein 17, 33
Specific Gravity 2.14
Solubility Information Solubility in water: practically insoluble
Formula Weight 241.93
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2, 5-Dibromothiophene
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dibromothiophene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.