missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromotoluene, 98%

Product Code. 11454017
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454017 25 g 25g
11444017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454017 Supplier Thermo Scientific Alfa Aesar Supplier No. A19418.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 615-59-8 | C7H6Br2 | 249.933 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-59-8
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000090
InChI Key QKEZTJYRBHOKHH-UHFFFAOYSA-N
Synonym 2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
PubChem CID 12006
IUPAC Name 1,4-dibromo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)Br

Specifications

Melting Point 5°C to 6°C
Density 1.813
Boiling Point 106°C to 108°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Aromatic
Refractive Index 1.602
Quantity 25 g
Beilstein 1859123
Formula Weight 249.94
Percent Purity 98%
Chemical Name or Material 2,5-Dibromotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.