missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromotoluene, 98%, Thermo Scientific Chemicals

Product Code. 10711711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10711711 25 g Glass bottle 25g
10687231 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10711711 Supplier Thermo Scientific Acros Supplier No. 263480250
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-59-8 | Molecular Formula: C7H6Br2 | Molecular Weight: 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 615-59-8
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00000090
InChI Key QKEZTJYRBHOKHH-UHFFFAOYSA-N
Synonym 2,5-dibromotoluene, benzene, 1,4-dibromo-2-methyl, toluene, 2,5-dibromo, 2,5-dibromo toluene, 3,6-dibromotoluene, 1,4-dibromo-2-methyl-benzene, pubchem3922, acmc-209mu2, chemwish ic06904
PubChem CID 12006
IUPAC Name 1,4-dibromo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Br)Br

Spezifikation

CAS Min % 98.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Colorless to Yellow
Density 1.8100g/mL
Boiling Point 135.0°C to 136.0°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3Br2
Refractive Index 1.6010 to 1.6030
Quantity 25 g
Beilstein 05, 308
Specific Gravity 1.81
Solubility Information Solubility in water: immiscible
Formula Weight 249.93
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 5-Dibromotoluene, 99%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 2,5-Dibromotoluene
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.