missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzotrifluoride, 98%

Product Code. 11433704
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433704 25 g 25g
11423704 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433704 Supplier Thermo Scientific Alfa Aesar Supplier No. A11310.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 320-50-3 | C7H3Cl2F3 | 214.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-50-3
Molecular Formula C7H3Cl2F3
Molecular Weight (g/mol) 214.996
MDL Number MFCD00000608
InChI Key DYBYUWVMLBBEMA-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzotrifluoride, 1,4-dichloro-2-trifluoromethyl benzene, benzene, 1,4-dichloro-2-trifluoromethyl, 2,5-dichloro-alpha,alpha,alpha-trifluorotoluene, 2,5-dichloro-a,a,a-trifluorotoluene, toluene, 2,5-dichloro-alpha,alpha,alpha-trifluoro-6ci,7ci,8ci, pubchem3450, ksc925c7r, 2,5-dichloro trifluoromethylbenzene
PubChem CID 35360
IUPAC Name 1,4-dichloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)Cl

Specifications

Density 1.483
Flash Point 76°C (168°F)
Refractive Index 1.483
Quantity 25 g
UN Number UN1760
Beilstein 1955442
Formula Weight 215
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.