missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 11353237
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353237 5 g 5g
11343237 1 g 1g
11363237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11353237 Supplier Thermo Scientific Alfa Aesar Supplier No. L13775.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 85482-13-9
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.921
MDL Number MFCD00236034
InChI Key UUVDOPTUDWJHFK-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzyl bromide, 2-bromomethyl-1,4-dichlorobenzene, 2,5-dichlorobenzylbromide, 1-bromomethyl-2,5-dichlorobenzene, alpha-bromo-2,5-dichlorotoluene, benzene, 2-bromomethyl-1,4-dichloro, 2-bromomethyl-1,4-dichloro-benzene, pubchem10208, acmc-1bjf0, 2,5-di-chloro-benzyl bromide
PubChem CID 3495744
IUPAC Name 2-(bromomethyl)-1,4-dichlorobenzene
SMILES C1=CC(=C(C=C1Cl)CBr)Cl

Spécification

Melting Point 39°C to 43°C
Quantity 5 g
UN Number UN3261
Beilstein 637803
Formula Weight 239.93
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzyl bromide
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.