missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzylamine, 97%, Thermo Scientific™

Product Code. 10273522
Change view
Click to view available options
:
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10273522 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10273522 Supplier Thermo Scientific Maybridge Supplier No. TL00182DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 10541-69-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H7Cl2N
Linear Formula Cl2C6H3CH2NH2
MDL Number MFCD00052391
Quantity 1g
Synonym 2,5-dichlorobenzylamine, 2,5-dichlorophenyl methanamine, benzenemethanamine, 2,5-dichloro, 2,5-dichloro-benzylamine, 1-2,5-dichlorophenyl methanamine, 2,5-dichlorophenyl methylamine, 2,5-dichlorobenzylam, acmc-1bxya, rarechem al bw 0384, 2,5-dichlorobenzyl amine
InChI Key AKGJLIXNRPNPCH-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)CN)Cl
IUPAC Name (2,5-dichlorophenyl)methanamine
Molecular Weight (g/mol) 176.04
PubChem CID 457600
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 2, 5-Dichlorobenzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.