missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophenol, 98%

Product Code. 11400293
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
Product Code. Quantity unitSize
11400293 5 g 5g
11484070 1 g 1g
11410293 25 g 25g
11420293 100 g 100g
4 options
This item is not returnable. View return policy

Product Code. 11400293

Brand: Thermo Scientific Alfa Aesar A11434.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5858-18-4 | C6H4Cl2S | 179.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5858-18-4
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00004831
InChI Key QIULLHZMZMGGFH-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophenol, 2,5-dichlorobenzene-1-thiol, benzenethiol, 2,5-dichloro, 2,5-dichloro thiophenol, thiophenol, 2,5-dichloro, 2,5-dichlorophenylmercaptan, pubchem6801, thiophenol,5-dichloro, 2,5-dichlorobenzenthiol, 2,5-dichloro-thiophenol
PubChem CID 79965
IUPAC Name 2,5-dichlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)S)Cl

Specifications

Melting Point 22°C to 25°C
Boiling Point 112°C to 116°C (50 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.619
Quantity 5 g
Beilstein 2042866
Sensitivity Air Sensitive
Formula Weight 179.07
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.