missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-methoxybenzaldehyde, Thermo Scientific™

Product Code. 10067689
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
10067689 1 g Amber Glass Bottle 1g
1 options
This item is not returnable. View return policy

Product Code. 10067689

Brand: Thermo Scientific Maybridge MAY00189DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 879093-08-0
Packaging Amber Glass Bottle
Molecular Formula C8H6F2O2
MDL Number MFCD06246882
Quantity 1 g
Synonym 2,5-difluoro-4-methoxy-benzaldehyde, 2,5-fluoro-4-methoxybenzaldehyde, pubchem17005, acmc-209qqk, 2,5-difluoro-4-methyoxybenzaldehyde, benzaldehyde,2,5-difluoro-4-methoxy, benzaldehyde, 2,5-difluoro-4-methoxy, 2,5-difluoro-4-methoxybenzaldehyde, 97+%
InChI Key DCGKDDVUUMOTDH-UHFFFAOYSA-N
SMILES COC1=C(F)C=C(C=O)C(F)=C1
IUPAC Name 2,5-difluoro-4-methoxybenzaldehyde
Molecular Weight (g/mol) 172.13
PubChem CID 3856938
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 2, 5-Difluoro-4-methoxybenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.