missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluoro-4-nitrobenzoic acid, 97%

Product Code. 15436617
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436617 5 g 5g
15426617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436617 Supplier Thermo Scientific Alfa Aesar Supplier No. H33757.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 116465-48-6 | C7H3F2NO4 | 203.101 g/mol

2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116465-48-6
Molecular Formula C7H3F2NO4
Molecular Weight (g/mol) 203.101
MDL Number MFCD03425705
InChI Key GPTNSBLYGCZJQV-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-nitrobenzoic acid, 2,5-difluoro-4-nitrobenzenecarboxylic acid, 2,5-difluoro-4-nitrobenzic acid, 2,5-difluoro-4-nitrobenzoicacid, 2,5-difluoro-4-nitro-benzoic acid, benzoic acid, 2,5-difluoro-4-nitro, 2,5-difluoro-4-nitrobenzenecarboxylicacid, pubchem9789, acmc-2099sr, ksc495m6l
PubChem CID 246375
SMILES C1=C(C(=CC(=C1F)[N+](=O)[O-])F)C(=O)O

Specifications

Melting Point 145°C to 147°C
Color Green to White
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 203.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Difluoro-4-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.