missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzyl bromide, 98%

Product Code. p-7025327
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11495616

Brand: Thermo Scientific Alfa Aesar B20752.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85117-99-3 | C7H5BrF2 | 207.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85117-99-3
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD00009897
InChI Key ONWGSWNHQZYCFK-UHFFFAOYSA-N
Synonym 2,5-difluorobenzyl bromide, 2-bromomethyl-1,4-difluorobenzene, 2,5-difluorobenzylbromide, alpha-bromo-2,5-difluorotoluene, benzene, 2-bromomethyl-1,4-difluoro, timtec-bb sbb006680, 2-bromomethyl-1,4-difluoro-benzene, pubchem10213, 2,2-difluorobenzylbromide, acmc-209q4i
PubChem CID 522830
IUPAC Name 2-(bromomethyl)-1,4-difluorobenzene
SMILES FC1=CC=C(F)C(CBr)=C1

Specifications

Density 1.609
Refractive Index 1.526
Quantity 10 g
UN Number UN3265
Beilstein 7089248
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material 2,5-Difluorobenzyl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.