missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenylacetonitrile, 97%, Thermo Scientific™

Product Code. 11499716
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11499716

Brand: Thermo Scientific Alfa Aesar B20246.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69584-87-8
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00009972
InChI Key UIMMFRUOZOWROM-UHFFFAOYSA-N
Synonym 2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
PubChem CID 592780
IUPAC Name 2-(2,5-difluorophenyl)acetonitrile
SMILES FC1=CC=C(F)C(CC#N)=C1

Specifications

Density 1.233
Boiling Point 90°C (5 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.4834
Quantity 5 g
UN Number UN3276
Beilstein 2576318
Formula Weight 153.13
Percent Purity 97%
Chemical Name or Material 2,5-Difluorophenylacetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.