missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorophenylacetonitrile, 97%, Thermo Scientific™

Product Code. 10763275 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10763275 5 g Glass bottle 5g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10763275

Brand: Thermo Scientific Acros 311660050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 69584-87-8
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00009972
InChI Key UIMMFRUOZOWROM-UHFFFAOYSA-N
Synonym 2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
PubChem CID 592780
IUPAC Name 2-(2,5-difluorophenyl)acetonitrile
SMILES FC1=CC=C(F)C(CC#N)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2300g/mL
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3CH2CN
Refractive Index 1.4824 to 1.4844
Quantity 5 g
Specific Gravity 1.23
Formula Weight 153.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 5-Difluorophenylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.