missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxyterephthalic acid, 97%

Product Code. p-7056129
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15462289

Brand: Thermo Scientific Alfa Aesar H64888.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610-92-4 | C8H6O6 | 198.13 g/mol

Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in hot dimethylformamide.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-92-4
Molecular Formula C8H6O6
Molecular Weight (g/mol) 198.13
MDL Number MFCD00132933
InChI Key OYFRNYNHAZOYNF-UHFFFAOYSA-N
Synonym 1,4-benzenedicarboxylic acid, 2,5-dihydroxy, 2,5-dihydroxyterephthalicacid, 2,5-dihydroxy-1,4-benzenedicarboxylic acid, 2,5-dihydroxybenzene-1,4-dicarboxylic acid, zlchem 699, acmc-1ayqi, 1, 2,5-dihydroxy, 2,5-dihydroxytelephthalic acid, #, 2,5-dihydroxyterephthalic acid
PubChem CID 69131
IUPAC Name 2,5-dihydroxyterephthalic acid
SMILES C1=C(C(=CC(=C1O)C(=O)O)O)C(=O)O

Specifications

Melting Point >300°C
Color Gray to Green
Quantity 5 g
Beilstein 2696052
Solubility Information Soluble in hot dimethylformamide.
Formula Weight 198.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Dihydroxyterephthalic acid
compliance-icons
Product Identifier
  • 2,5-Dihydroxyterephthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,5-Dihydroxyterephthalic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.