missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyaniline, 99%

Product Code. 10724081 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10724081 250 g Glass bottle 250g
10437772 5 g Glass bottle 5g
10225910 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10724081 Supplier Thermo Scientific Acros Supplier No. 150852500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 102-56-7 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-56-7
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008368
InChI Key NAZDVUBIEPVUKE-UHFFFAOYSA-N
Synonym benzenamine, 2,5-dimethoxy, aminohydroquinone dimethyl ether, 1-amino-2,5-dimethoxybenzene, 2,5-dimethoxybenzenamine, aniline, 2,5-dimethoxy, unii-v3z5u3fl10, 2,5 dimethoxyaniline, 2,5-dimethoxyphenylamine, dimethoxyaniline 2,5-, 2,5-dimethoxy aniline
PubChem CID 7613
IUPAC Name 2,5-dimethoxyaniline
SMILES COC1=CC=C(OC)C(N)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 79.0°C to 82.0°C
Color Brown to Gray
Boiling Point 140.0°C (7.0 mmHg)
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3NH2
Quantity 250 g
Beilstein 13, 788
Solubility Information Solubility in water: 4g/L (40°C). Other solubilities: soluble in most common organic solvents
Formula Weight 153.18
Percent Purity 99%
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 2, 5-Dimethoxyaniline, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.