missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxycinnamic acid, predominantly trans, 99%, Thermo Scientific™

Product Code. 11410217
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410217 5 g 5g
11420217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410217 Supplier Thermo Scientific Alfa Aesar Supplier No. B21649.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10538-51-9
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00004378
InChI Key JPQWWJZORKTMIZ-ZZXKWVIFSA-N
Synonym 2,5-dimethoxycinnamic acid, 3-2,5-dimethoxyphenyl acrylic acid, e-3-2,5-dimethoxyphenyl acrylic acid, 2e-3-2,5-dimethoxyphenyl acrylic acid, 3-2,5-dimethoxyphenyl-2-propenoic acid, 2e-3-2,5-dimethoxyphenyl prop-2-enoic acid, 3-2,5-dimethoxyphenyl prop-2-enoic acid, rarechem bk hc t324, timtec-bb sbb000471, trans-2,5-dimethoxycinnamic acid
PubChem CID 697594
IUPAC Name (E)-3-(2,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=C(C=C1)OC)C=CC(=O)O

Specifications

Melting Point 147°C to 150°C
Quantity 5 g
Formula Weight 208.21
Percent Purity 99%
Chemical Name or Material 2,5-Dimethoxycinnamic acid, predominantly trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.