missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylcinnamic acid, 98+%, Thermo Scientific™

Product Code. 11381407
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381407 10 g 10g
11371407 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381407 Supplier Thermo Scientific Alfa Aesar Supplier No. L01799.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,5-Dimethylcinnamic acid, is used for organic synthesize intermediate

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95883-10-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00014377
InChI Key FAIBVLSPNAHVSQ-UHFFFAOYSA-N
Synonym 3-2,5-dimethylphenyl prop-2-enoic acid, 2,5-dimethylcinnamic acid, 2-propenoic acid, 3-2,5-dimethylphenyl
PubChem CID 53401582
IUPAC Name 3-(2,5-dimethylphenyl)prop-2-enoic acid
SMILES CC1=CC(C=CC(O)=O)=C(C)C=C1

Specifications

Melting Point 129°C to 131°C
Quantity 10 g
Beilstein 3244516
Solubility Information Insoluble in water.
Formula Weight 176.22
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethylcinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.