missing translation for 'onlineSavingsMsg'
Learn More

2,5-dimethylphenyl isothiocyanate, Thermo Scientific™

Product Code. 10648755
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10648755 5 g 5g
10681365 10 g 10g
10337164 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10648755 Supplier Thermo Scientific Maybridge Supplier No. SB01618DE
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 19241-15-7
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
InChI Key RCBXZRJVMSWGAO-UHFFFAOYSA-N
PubChem CID 140499
IUPAC Name 2-isothiocyanato-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)N=C=S

Specifications

Quantity 5 g
Packaging Amber glass bottle
Assay Percent Range 97%
Synonym 2,5-dimethylphenyl isothiocyanate, benzene,2-isothiocyanato-1,4-dimethyl, 2,5-dimethylbenzenisothiocyanate, 2-isothiocyanato-1,4-dimethyl-benzene, 2,5-xylyl isothiocyanate, 2,5-dimethylphenyl-isothiocyanate, 2,5-dimethylphenylisothiocyanate, #, benzene, 2-isothiocyanato-1,4-dimethyl
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.