missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10433510 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10433510

Brand: Thermo Scientific Acros 359260250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 85199-06-0
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD01863525
InChI Key OOMZKLJLVGQZGV-UHFFFAOYSA-N
Synonym 2,5-dimethylphenyl boronic acid, 2,5-dimethylbenzeneboronic acid, 2,5-dimethylphenylboronicacid, 2,5-dimethylphenyl boranediol, p-xylene-2-boronic acid, boronic acid, 2,5-dimethylphenyl, pubchem1831, p-xyleneboronic acid, ksc448a1r, 2,5-dimehylphenylboronic acid
PubChem CID 2734347
IUPAC Name (2,5-dimethylphenyl)boronic acid
SMILES CC1=CC=C(C)C(=C1)B(O)O

Specifications

Melting Point 186.0°C to 191.0°C
CAS Min % 96.0
CAS Max % 100.0
Quantity 25 g
Packaging Glass bottle
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 149.98
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylphenylboronic acid
compliance-icons
Product Identifier
  • 2,5-Dimethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.