accessibility menu, dialog, popup

UserName

CAS RN 85199-06-0

CH3CH3BHOOH

CAS RN 85199-06-0

IUPAC Name: (2,5-dimethylphenyl)boronic acid
Synonyms: 2,5-dimethylphenylboronic acid
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: OOMZKLJLVGQZGV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(=C1)B(O)O
Quantity 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)