missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenylhydrazine hydrochloride, 98%, Thermo Scientific™

Product Code. 11373186
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373186 100 g 100g
11353186 5 g 5g
11363186 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11373186 Supplier Thermo Scientific Alfa Aesar Supplier No. L12496.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56737-78-1
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00013382
InChI Key OMOFMSZLYCEIAF-UHFFFAOYSA-N
Synonym 2,5-dimethylphenylhydrazine hydrochloride, 2,5-dimethylphenyl hydrazine hydrochloride, 2,5-dimethylphenylhydrazinehydrochloride, 2,3-dimethylphenylhydrazinehydrochloride, pubchem6400, acmc-1bf0t, ksc491e8b, 2-hydrazino-m-xylene hydrochloride, 2,5-dimethyl-phenylhydrazine hydrochloride, 1-2,5-dimethylphenyl hydrazine hydrochloride
PubChem CID 2734058
IUPAC Name (2,5-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=CC(=C(C=C1)C)NN.Cl

Specifications

Melting Point ∽205°C (decomposition)
Quantity 100 g
Beilstein 6118180
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylphenylhydrazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.