missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASCT2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13444699
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13444699 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13444699 Supplier Invitrogen™ Supplier No. PA533029
  • ONLINE EXCLUSIVE PRICE
    530.00 €
    376.30 € / 50µg
    Save 153.70 €
    29% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Marmoset (100%) Monkey (94%) Dog, Horse (82%).

SLC1A5 (Solute carrier family 1 (neutral amino acid transporter), member 5) is a member of the Na(+)-dependent amino acid transporter superfamily. It has a broad substrate specificity, a preference for zwitterionic amino acids, and a sodium-dependence. It accepts as substrates all neutral amino acids, including glutamine, asparagine, and branched-chain and aromatic amino acids, and excludes methylated amino acids, anionic amino acids, and cationic amino acids. It acts as a cell surface receptor for feline endogenous virus RD114, baboon M7 endogenous virus and type D simian retroviruses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SLC1A5
Gene Accession No. Q15758
Gene Alias AAAT; ASC-like Na(+)-dependent neutral amino acid transporter ASCT2; Asct2; ATB(0); ATB0; ATBO; baboon M7 virus receptor; CAZ-associated structural protein; H4-ASCT2; H4-system ASC-like transporter; insulin-activated amino acid transporter; M7V1; M7VS1; neutral amino acid transporter B; neutral amino acid transporter B(0); R16; RD114 virus receptor; RD114/simian type D retrovirus receptor; RDR; RDRC; SLC1A5; Slc1a7; sodium-dependent neutral amino acid transporter ASCT2; sodium-dependent neutral amino acid transporter type 2; solute carrier family 1 (neutral amino acid transporter), member 5; solute carrier family 1 member 5; solute carrier family 1, member 7
Gene Symbols SLC1A5
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human SLC1A5.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6510
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.