missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(hydroxymethyl)-p-cresol, tech. 90%

Product Code. 11493607
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493607 25 g 25g
11403617 100 g 100g
11413617 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493607 Supplier Thermo Scientific Alfa Aesar Supplier No. A17369.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-04-3 | C9H12O3 | 168.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-04-3
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004619
InChI Key KUMMBDBTERQYCG-UHFFFAOYSA-N
Synonym 2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene
PubChem CID 7039
IUPAC Name 2,6-bis(hydroxymethyl)-4-methylphenol
SMILES CC1=CC(=C(C(=C1)CO)O)CO

Specifications

Melting Point 119°C to 126°C
Quantity 25 g
Beilstein 1240237
Formula Weight 168.19
Percent Purity 90%
Grade Technical
Chemical Name or Material 2,6-Bis(hydroxymethyl)-p-cresol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.