missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)benzoic acid, 98%

Product Code. 11474477
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474477 5 g 5g
11464477 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474477 Supplier Thermo Scientific Alfa Aesar Supplier No. B20486.06
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24821-22-5 | C9H4F6O2 | 258.119 g/mol

2,6-Bis(trifluoromethyl)benzoic acid is used as an organic intermediate in synthesis. It is also used as a pharmaceutical intermediate and organic intermediate in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzoic acid is used as an organic intermediate in synthesis. It is also used as a pharmaceutical intermediate and organic intermediate in chemical synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24821-22-5
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.119
MDL Number MFCD00000376
InChI Key XZNLSDPNMNWCRE-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl benzoic acid, benzoic acid, 2,6-bis trifluoromethyl, 2,6-bis-trifluoromethylbenzoic acid, 2,6-di trifluoromethyl benzoic acid, 2,6-bis-trifluoromethyl-benzoic acid, 2,6-bis trifluoromethyl-benzoic acid, pubchem4988, acmc-1co7v, rarechem al bo 0513, timtec-bb sbb000962
PubChem CID 90618
IUPAC Name 2,6-bis(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C(=C1)C(F)(F)F)C(=O)O)C(F)(F)F

Specifications

Melting Point 136°C to 140°C
Odor Odorless
Quantity 5 g
Beilstein 2057459
Solubility Information Soluble in water.
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 2,6-Bis(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.