missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)benzoic acid, 98%

Product Code. 10189629 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10189629 1 g Glass bottle 1g
10129399 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10189629 Supplier Thermo Scientific Acros Supplier No. 214460010
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24821-22-5 | C9H4F6O2 | 258.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24821-22-5
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.11
InChI Key XZNLSDPNMNWCRE-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl benzoic acid, benzoic acid, 2,6-bis trifluoromethyl, 2,6-bis-trifluoromethylbenzoic acid, 2,6-di trifluoromethyl benzoic acid, 2,6-bis-trifluoromethyl-benzoic acid, 2,6-bis trifluoromethyl-benzoic acid, pubchem4988, acmc-1co7v, rarechem al bo 0513, timtec-bb sbb000962
PubChem CID 90618
IUPAC Name 2,6-bis(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C(=C1)C(F)(F)F)C(=O)O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 137.7°C to 138.6°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CO2H
Quantity 1 g
Formula Weight 258.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Bis(trifluoromethyl)benzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.