missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylphenol, 99.8%

Product Code. 13206319 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13206319 5 kg Plastic drum 5kg
10378590 50 g Plastic bottle 50g
10296460 250 g Plastic bottle 250g
13286309 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13206319 Supplier Thermo Scientific Acros Supplier No. 219830050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 128-37-0 | C15H24O | 220.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.35
MDL Number MFCD00011644
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

CAS Min % 99.75
CAS Max % 100.0
Melting Point 69.0°C to 71.0°C
Color White
Boiling Point 265.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 99.8% min. (GC)
Packaging Plastic drum
Linear Formula [(CH3)3C]2C6H2(CH3)OH
Quantity 5 kg
Beilstein 06,III,2073
Fieser 15,113
Merck Index 15,155
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in toluene,soluble in methanol,ethanol,isopropanol,acetone,benzene,methyl ethyl ketone,petroleum ether,,cellowolve and most other hydrocarbon solvents
Viscosity 3.47 mm2/s (0°C)
Formula Weight 220.35
Percent Purity 99.8%
Physical Form Crystals
Chemical Name or Material 2, 6-Di-tert-butyl-4-methylphenol, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Di-tert-butyl-4-methylphenol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste d.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.