missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-p-cresol, TRC

Product Code. 30009901
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009901 10 g 10g
17912605 500 mg 500mg
17954225 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009901 Supplier Toronto Research Chemicals Supplier No. TRCD42948010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15 H24 O
Molecular Weight (g/mol) 220.35
Synonym p-Cresol, 2,6-di-tert-butyl- (7CI,8CI), 2,6-Bis(1,1-dimethylethyl)-4-methylphenol, 2,6-Bis(tert-butyl)-4-methylphenol, 2,6-Di(tert-butyl)hydroxytoluene, 2,6-Di-t-butyl-4-methylphenol, 2,6-Di-tert-butyl-4-cresol, 2,6-Di-tert-butyl-4-hydroxytoluene, 2,6-Di-tert-butyl-4-methyl phenol, 2,6-Di-tert-butyl-4-methyl-1-hydroxybenzene, 2,6-Di-tert-butyl-4-methylhydroxybenzene, 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-p-cresole, 2,6-Di-tert-butyl-p-methylphenol, 2,6-Di-tert-butylcresol, 2,6-Di-tert-butylmethylphenol, 2,6-Ditertbutyl paracresol, 2,6-tert-Butyl-4-methylphenol, 3,5-Bis(1,1-dimethylethyl)-4-hydroxytoluene, 3,5-Di-tert-butyl-4-hydroxytoluene, 3,5-Di-tert-butyl-p-hydroxytoluene, 4-Hydroxy-3,5-di-tert-butyltoluene, 4-Methyl-2,6-bis(1,1-dimethylethyl)phenol, 4-Methyl-2,6-di-tert-butylphenol, 4-Methyl-2,6-ditertbutyl phenol, AO 29, AO 4, AO 4K, AOX 4, AOX 4K, Addiox 1005, Additin RC 1220, Advastab 401, Agidol, Agidol 1, Agidol 1A, Alkofen BP, Antage BHT, Antioxidant 246, Antioxidant 264, Antioxidant 29, Antioxidant 30, Antioxidant 4, Antioxidant 4K, Antioxidant 501, Antioxidant BHT, Antioxidant DBPC, Antioxidant KB, Antioxidant MPJ, Antioxidant OC, Antioxidant T 501, Antox QT, B-NOX BHT-P, BAT, BHT, BHT 264, BHT 501, BHT Swanox, BHT-C, Buks, Butylated hydroxytoluene, CAO 1, CAO 3, Catalin CAO 3, Chemanox 11, DBPC, Dalpac, Deenax, Di-tert-Butyl-4-methylphenol, Di-tert-butyl-p-cresol, Di-tert-butylcresol, Dibunol, Dibutyl paracresol, Dibutylated hydroxytoluene, Dibutylcresol, Dibutylhydroxytoluene, E 321, FG, Grindox BHT, H-BHT, Impruvol, Ionol, Ionol 1, Ionol 330, Ionol BHT, Ionol CP, Ionol K, Ionole, K-NOX-BHT, Kerabit, Lavinix BHT, Lowinox BHT, Lubrizol 817, M 4801, NK-BHT, NSC 6347, Na-Lube AO 220, Naugard BHT, Nipanox, Nipanox BHT, Nocrac 200, Nocrac 2000, Nonox TBC, P 21, P 21 (phenol), PN 01, Parabar 441, Permanax BHT, Popol, Ralox BHT, Selosol H 633, Stavox, Sumilizer BHT, Sumilizer BHT-R, Sustane BHT, Swanox BHT, T 501, T 501 (phenol), T 502A, Tenamene 3, Tenox BHT, Tonarol, Topanol, Topanol BHT, Topanol C, Topanol O, Topanol O-FG, Topanol OC, Topanol OL, Ultranox 226, Vanlube PC, Vanlube PCX, Vanox PC, Vianol, Vulkanox BHT, Vulkanox KB, Yoshinox BHT, o-Di-tert-butyl-p-methylphenol
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3
Formula Weight 220.1827
Percent Purity >95%
Chemical Name or Material Butylhydroxytoluene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.