missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butylphenol, 98%

Product Code. 11413477
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413477 250 g 250g
11403477 1000 g 1000g
11423477 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413477 Supplier Thermo Scientific Alfa Aesar Supplier No. A17244.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 128-39-2 | C14H22O | 206.33 g/mol

Widely used as antioxidant in fuels, lubricants and polymers. Employed as a synthetic intermediate for the production of higher molecular weight phenolic antioxidants. Used as an oxidation inhibitor and stabilizer (e.g. for fuel, oil and gasoline) and also used in plastics and rubber. Also applied as an intermediate and an antioxidant in aviation gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Widely used as antioxidant in fuels, lubricants and polymers. Employed as a synthetic intermediate for the production of higher molecular weight phenolic antioxidants. Used as an oxidation inhibitor and stabilizer (e.g. for fuel, oil and gasoline) and also used in plastics and rubber. Also applied as an intermediate and an antioxidant in aviation gasoline.

Solubility
Soluble in water at 20°C 0.003g/L. Solubility in methanol is almost transparent.

Notes
Store away from strong oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-39-2
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.33
MDL Number MFCD00008820
InChI Key DKCPKDPYUFEZCP-UHFFFAOYSA-N
Synonym 2,6-di-tert-butylphenol, 2,6-di-t-butylphenol, 2,6-bis tert-butyl phenol, 2,6-bis 1,1-dimethylethyl phenol, phenol, 2,6-bis 1,1-dimethylethyl, 2,6 di-tert-butylphenol, ethanox 701, isonox 103, ethyl 701, ethyl an 701
PubChem CID 31405
IUPAC Name 2,6-ditert-butylphenol
SMILES CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C

Specifications

Melting Point 34°C to 39°C
Density 0.914
Boiling Point 252°C to 253°C
Flash Point 120°C (248°F)
Odor Phenol-like
Quantity 250 g
UN Number UN3077
Beilstein 1841887
Solubility Information Soluble in water at 20°C 0.003g/L. Solubility in methanol is almost transparent.
Formula Weight 206.33
Percent Purity 98%
Chemical Name or Material 2,6-Di-tert-butylphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Di-tert-butylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2,6-Di-tert-butylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.