missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butylpyridine, 97+%

Product Code. 15412108
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412108 25 g 25g
15492098 1 g 1g
15402108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412108 Supplier Thermo Scientific Alfa Aesar Supplier No. H55761.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 585-48-8 | C13H21N | 191.318 g/mol

2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.

Solubility
Miscible with alcohol, acetone, and hexane. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-48-8
Molecular Formula C13H21N
Molecular Weight (g/mol) 191.318
MDL Number MFCD00006306
InChI Key UWKQJZCTQGMHKD-UHFFFAOYSA-N
Synonym 2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119
PubChem CID 68510
IUPAC Name 2,6-ditert-butylpyridine
SMILES CC(C)(C)C1=NC(=CC=C1)C(C)(C)C

Specifications

Density 0.867
Boiling Point 100°C to 101°C (23 mmHg)
Flash Point 71°C (160°F)
Refractive Index 1.473
Quantity 25 g
Beilstein 125886
Merck Index 14,3036
Solubility Information Miscible with alcohol,acetone,and hexane. Immiscible with water.
Formula Weight 191.32
Percent Purity ≥97%
Chemical Name or Material 2,6-Di-tert-butylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.