missing translation for 'onlineSavingsMsg'
Learn More

2,6-dibromo-3-chloro-4-fluoroaniline, Thermo Scientific™

Product Code. p-4391289
Click to view available options
Quantity:
25 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
50g
This item is not returnable. View return policy

Product Code. 10021424

Brand: Thermo Scientific Maybridge BTB04528EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-09-8
Molecular Formula C6H3Br2ClFN
Molecular Weight (g/mol) 303.353
InChI Key PRZBEZGEEPNKRO-UHFFFAOYSA-N
PubChem CID 2736480
IUPAC Name 2,6-dibromo-3-chloro-4-fluoroaniline
SMILES C1=C(C(=C(C(=C1Br)N)Br)Cl)F

Specifications

Packaging Amber Glass Bottle
Quantity 25 g
Synonym 3-chloro-2,6-dibromo-4-fluoroaniline, 2,6-dibromo-3-chloro-4-fluorobenzenamine, 1-chloro-2,6-dibromo-4-fluoroaniline, benzenamine, 2,6-dibromo-3-chloro-4-fluoro, benzenamine,2,6-dibromo-3-chloro-4-fluoro, pubchem2948, acmc-1c6hf, 2,6-dibromo-3-chloro-4-fluorophenylamine
Formula Weight 303.36
Percent Purity 97%
Chemical Name or Material 2, 6-dibromo-3-chloro-4-fluoroaniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.