missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-fluorophenol, 98+%, Thermo Scientific™

Product Code. 11496516
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11496516

Brand: Thermo Scientific Alfa Aesar B24782.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-20-7
Molecular Formula C6H3Br2FO
Molecular Weight (g/mol) 269.895
MDL Number MFCD00002317
InChI Key RRAZCUUOWIDAJS-UHFFFAOYSA-N
Synonym phenol, 2,6-dibromo-4-fluoro, 2,6-dibromo-4-fluoro-phenol, pubchem2868, acmc-209i7j, ksc494q8h, emolecules 492569, 1-bromo-3,4,5-trifluorobezene, attercop-chm at113643, 2,6-dibromo-4-fluorophenol
PubChem CID 67654
IUPAC Name 2,6-dibromo-4-fluorophenol
SMILES C1=C(C=C(C(=C1Br)O)Br)F

Specifications

Melting Point 54°C to 56°C
Odor Phenol-like
Quantity 25 g
Beilstein 1868033
Formula Weight 269.91
Percent Purity ≥98%
Chemical Name or Material 2,6-Dibromo-4-fluorophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.