missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-(trifluoromethyl)aniline, 97%

Product Code. 11392926
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392926 5 g 5g
11382926 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392926 Supplier Thermo Scientific Alfa Aesar Supplier No. L10658.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 72678-19-4 | C7H4Br2F3N | 318.919 g/mol

Used in enantio-and diastereoselective addition reation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in enantio-and diastereoselective addition reation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72678-19-4
Molecular Formula C7H4Br2F3N
Molecular Weight (g/mol) 318.919
MDL Number MFCD00068181
InChI Key DRSMEHXBOXHXDX-UHFFFAOYSA-N
Synonym 2,6-dibromo-4-trifluoromethyl aniline, 4-amino-3,5-dibromobenzotrifluoride, 2,6-dibromo-4-trifluoromethyl phenylamine, benzenamine,2,6-dibromo-4-trifluoromethyl, benzenamine, 2,6-dibromo-4-trifluoromethyl, pubchem2805, acmc-209ony, buttpark 29\06-99, 2,6-dibromo-4-trifluoromethyl-phenylamine
PubChem CID 144565
IUPAC Name 2,6-dibromo-4-(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1Br)N)Br)C(F)(F)F

Specifications

Melting Point 36°C to 40°C
Boiling Point 64°C to 65°C (0.1 mmHg)
Quantity 5 g
UN Number UN2811
Beilstein 6314196
Solubility Information Insoluble in water.
Formula Weight 318.93
Percent Purity 97%
Chemical Name or Material 2,6-Dibromo-4-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.