missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromoaniline, 97%

Product Code. 11334534
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334534 2 g 2g
11344534 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Artikelnummer. 11334534 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. L01285.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 608-30-0 | C6H5Br2N | 250.92 g/mol

2,5-Dibromoaniline was used in the synthesis of 2,4,5-tribromoaniline1. It was also used as starting reagent in the regioselective synthesis of 2-(3,4-dimethoxyphenyl)-5-bromobenzothiazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromoaniline was used in the synthesis of 2,4,5-tribromoaniline1. It was also used as starting reagent in the regioselective synthesis of 2-(3,4-dimethoxyphenyl)-5-bromobenzothiazole.

Solubility
Insoluble in water.

Notes
Incompatible with acids, Acid chlorides, Acid anhydrides, Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 608-30-0
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.92
MDL Number MFCD00007638
InChI Key XIRRDAWDNHRRLB-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dibromo, 2,6-dibromophenylamine, 2,6-dibromobenzenamine, aniline, 2,6-dibromo, 2,6-dibromo-phenylamine, 2,6-dibromo-aniline, acmc-1aycf, 2,6-dibromoaniline, 2,6-dibromo-phenyl-amine, ksc356c3l
PubChem CID 69098
IUPAC Name 2,6-dibromoaniline
SMILES NC1=C(Br)C=CC=C1Br

Spezifikation

Melting Point 79°C to 83°C
Boiling Point 262°C to 264°C
Odor Characteristic
Quantity 2 g
UN Number UN2811
Beilstein 2207094
Solubility Information Insoluble in water.
Formula Weight 250.93
Percent Purity 97%
Chemical Name or Material 2,6-Dibromoaniline

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.