missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromoaniline, 97%

Product Code. 11344534
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344534 10 g 10g
11334534 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344534 Supplier Thermo Scientific Alfa Aesar Supplier No. L01285.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-30-0 | C6H5Br2N | 250.92 g/mol

2,5-Dibromoaniline was used in the synthesis of 2,4,5-tribromoaniline1. It was also used as starting reagent in the regioselective synthesis of 2-(3,4-dimethoxyphenyl)-5-bromobenzothiazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromoaniline was used in the synthesis of 2,4,5-tribromoaniline1. It was also used as starting reagent in the regioselective synthesis of 2-(3,4-dimethoxyphenyl)-5-bromobenzothiazole.

Solubility
Insoluble in water.

Notes
Incompatible with acids, Acid chlorides, Acid anhydrides, Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-30-0
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.92
MDL Number MFCD00007638
InChI Key XIRRDAWDNHRRLB-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dibromo, 2,6-dibromophenylamine, 2,6-dibromobenzenamine, aniline, 2,6-dibromo, 2,6-dibromo-phenylamine, 2,6-dibromo-aniline, acmc-1aycf, 2,6-dibromoaniline, 2,6-dibromo-phenyl-amine, ksc356c3l
PubChem CID 69098
IUPAC Name 2,6-dibromoaniline
SMILES NC1=C(Br)C=CC=C1Br

Specifications

Melting Point 79°C to 83°C
Boiling Point 262°C to 264°C
Odor Characteristic
Quantity 10 g
UN Number UN2811
Beilstein 2207094
Solubility Information Insoluble in water.
Formula Weight 250.93
Percent Purity 97%
Chemical Name or Material 2,6-Dibromoaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.