missing translation for 'onlineSavingsMsg'
Learn More

2,6-dichloro-1,3-benzothiazole, Thermo Scientific™

Product Code. 10130792
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10130792 1 g 1g
10446862 10 g 10g
10644585 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10130792 Supplier Thermo Scientific Maybridge Supplier No. SB01025DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3622-23-9
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.068
InChI Key QDZGJGWDGLHVNK-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzothiazole, 2,6-dichlorobenzo d thiazole, benzothiazole, 2,6-dichloro, 2,6-dichloro-benzothiazole, 2,6-dichloro benzothiazole, pubchem17324, acmc-1adss, ksc490o9j
PubChem CID 77176
IUPAC Name 2,6-dichloro-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Cl)SC(=N2)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-1, 3-benzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.